Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ILC Dover Breathing Tube Cover Installation Tube

Catalog No. 19631111
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-631-111 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-631-111 Supplier ILC Dover Supplier No. 60006150801

missing translation for 'validMainframeMsg'

Save time by using this tool to easily slip the cover onto the breathing tube to help prevent contamination.

  • Helps prevent contamination

Specifications

Color White
Product Type Breathing Tube Installation Accessory
For Use With (Application) Installation of breathing tube
For Use With (Equipment) PAPR Systems
Model 6000-61508-01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.