Learn More
Enterprise Technology Solutions Brecknell Portable Shipping Scale, LCD Display, 400lb, 15"x12", White
SDP
Catalog No. 5000829287
Encompass_Preferred
Portable shipping scale is well-suited for shipping or warehouse applications and general-purpose weighing with computer interface. The scale accurately weighs up to 400 lb. and is compatible with UPS Shipping software. Scale includes four AAA batteries. A large, easy-to-read, 0.65" LCD screen can be wall-mounted or desk-mounted for easy visibility. The large keyboard switches work with gloved hands. This four-load cell design provides weight accurate for all major carrier shipments. Scale includes an RS232 link that is compatible with most carrier-provided shipping systems as well as a PC Serial Interface Cable.
Catalog No. 50-008-29287 Supplier Enterprise Technology Solutions Supplier No. SBWLPS400
Portable shipping scale is well-suited for shipping or warehouse applications and general-purpose weighing with computer interface. The scale accurately weighs up to 400 lb. and is compatible with UPS Shipping software. Scale includes four AAA batteries. A large, easy-to-read, 0.65" LCD screen can be wall-mounted or desk-mounted for easy visibility. The large keyboard switches work with gloved hands. This four-load cell design provides weight accurate for all major carrier shipments. Scale includes an RS232 link that is compatible with most carrier-provided shipping systems as well as a PC Serial Interface Cable.