Learn More
Enterprise Technology Solutions Brecknell Scale,Balance,LCD,500g Cap, 4"x5-1/2"x4/5",SR/BK
SDP
Catalog No. 5000829290
Encompass_Preferred
The EPB Balance scale is convenient, compact and portable. This elegant-looking scale offers unparalleled weighing performance. For weighing jewelry, food ingredients and other small, lightweight objects, the EPB is the perfect fit. It features an ABS case with stainless steel weight platter and an easy-to-read, white backlit display that is ideal for low-light environments. It includes a plastic protector cover for the platter and two AAA batteries. Auto-off function shuts off after 60 seconds to conserve battery life. Other features include an overload and low-battery indicator as well as a push-button tare. Functions include weight mode, tare, counting and calibration. Units include kg, g, lb, oz, tl and ct.
Catalog No. 50-008-29290 Supplier Enterprise Technology Solutions Supplier No. SBWEPB500
The EPB Balance scale is convenient, compact and portable. This elegant-looking scale offers unparalleled weighing performance. For weighing jewelry, food ingredients and other small, lightweight objects, the EPB is the perfect fit. It features an ABS case with stainless steel weight platter and an easy-to-read, white backlit display that is ideal for low-light environments. It includes a plastic protector cover for the platter and two AAA batteries. Auto-off function shuts off after 60 seconds to conserve battery life. Other features include an overload and low-battery indicator as well as a push-button tare. Functions include weight mode, tare, counting and calibration. Units include kg, g, lb, oz, tl and ct.