Learn More
Enterprise Technology Solutions Brecknell Scale,Hanging,Digital,Handheld,55 lb Cap,GY
SDP
Catalog No. 5000829286
Encompass_Preferred
Modern, ElectroSamson Handheld Digital Scale offers all the qualities of a traditional hanging balance with the simplicity, convenience and accuracy of digital operation. Large LCD is easy to read. Convenient hold feature allows one-person operation on heavier loads. Angled display lets you operate with only one hand. Scale features a strong, aluminum grip handle and memory tare function that retains tare even when powered off. Weight capacity is 55 lb.
Catalog No. 50-008-29286 Supplier Enterprise Technology Solutions Supplier No. SBWES55
Modern, ElectroSamson Handheld Digital Scale offers all the qualities of a traditional hanging balance with the simplicity, convenience and accuracy of digital operation. Large LCD is easy to read. Convenient hold feature allows one-person operation on heavier loads. Angled display lets you operate with only one hand. Scale features a strong, aluminum grip handle and memory tare function that retains tare even when powered off. Weight capacity is 55 lb.