Learn More
Medchemexpress LLC HY-106048 25mg Medchemexpress, Bredinin aglycone CAS:56973-26-3 Purity:>98%
SDP
Catalog No. 501968312
Encompass_Preferred
Medchemexpress, HY-106048 25mg Bredinin aglycone CAS:56973-26-3 Bredinin aglycone (5-Hydroxy-1H-imidazole-4-carboxamide) is a purine nucleotide analogue. Bredinin aglycone can be used to examine the efficiency of catalysts for the preparation of purine nucleotide analogues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8312 Supplier Medchemexpress LLC Supplier No. HY10604825MG
Medchemexpress, HY-106048 25mg Bredinin aglycone CAS:56973-26-3 Bredinin aglycone (5-Hydroxy-1H-imidazole-4-carboxamide) is a purine nucleotide analogue. Bredinin aglycone can be used to examine the efficiency of catalysts for the preparation of purine nucleotide analogues. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.