Learn More
Selleck Chemical LLC Brefeldin A 100mg 20350-15-6
SDP
Supplier:  Selleck Chemical LLC S7046100MG
Encompass_Preferred
Brefeldin A is a lactone antibiotic and ATPase inhibitor for protein transport with IC50 of 0.2 ?M in HCT 116 cells, induces cancer cell differentiation and apoptosis. It could also improve the HDR(homology-directed repair) efficiency and be an enhancer of CRISPR-mediated HDR. Brefeldin A is also an inhibitor of autophagy and mitophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4350