Learn More
Apexbio Technology LLC Brefeldin A(Synonyms: BFA, Ascotoxin, Cyanein, Decumbin, Brefeldin, γ), 10mM (in 1mL DMSO), CAS: 20350-15-6.
SDP
Catalog No. 5000060142
Encompass_Preferred
Brefeldin A (CAS 20350-15-6) commonly abbreviated as BFA is a small-molecule inhibitor targeting ATPase activity with a reported IC50 of approximately 0 2 M Its primary mechanism of action involves disruption of intracellular vesicle transport by blocking protein trafficking from the endoplasmic reticulum (ER) to the Golgi apparatus and inhibiting GTP/GDP exchange Through these activities BFA reduces ATP-mediated vesicular exocytosis thereby diminishing stimulus-dependent hyperalgesia Additionally it induces endoplasmic reticulum stress and promotes p53 expression in tumor cell models like MCF-7 and HeLa ultimately enhancing apoptosis in colorectal cancer cells (HCT116) BFA is widely employed as a pharmacological tool in cellular biology research to study protein secretion vesicular transport dynamics and ER stress pathways
Catalog No. 50-000-60142 Supplier Apexbio Technology LLC Supplier No. B14005.1
Brefeldin A (CAS 20350-15-6) commonly abbreviated as BFA is a small-molecule inhibitor targeting ATPase activity with a reported IC50 of approximately 0 2 M Its primary mechanism of action involves disruption of intracellular vesicle transport by blocking protein trafficking from the endoplasmic reticulum (ER) to the Golgi apparatus and inhibiting GTP/GDP exchange Through these activities BFA reduces ATP-mediated vesicular exocytosis thereby diminishing stimulus-dependent hyperalgesia Additionally it induces endoplasmic reticulum stress and promotes p53 expression in tumor cell models like MCF-7 and HeLa ultimately enhancing apoptosis in colorectal cancer cells (HCT116) BFA is widely employed as a pharmacological tool in cellular biology research to study protein secretion vesicular transport dynamics and ER stress pathways