Learn More
AdipoGen (+)-Brefeldin A,Chemical. CAS: 20350-15-6
SDP
Catalog No. 50464324
Encompass_Preferred
(+)-Brefeldin A, AdipoGen Life Sciences. Chemical. CAS: 20350-15-6. Formula: C16H24O4. MW: 280.4. Isolated from Penicillium janthinellum. Protein transport from ER to Golgi inhibitor. ADP-ribosylation factor (Arf) inhibitor. Cytotoxic. Antibiotic. Antiviral. Apoptosis inducer. Antitumor compound. Intracellular collagen degradation inhibitor. Reviews.
Catalog No. 50-464-324 Supplier AdipoGen Supplier No. AGCN20018M025
(+)-Brefeldin A, AdipoGen Life Sciences. Chemical. CAS: 20350-15-6. Formula: C16H24O4. MW: 280.4. Isolated from Penicillium janthinellum. Protein transport from ER to Golgi inhibitor. ADP-ribosylation factor (Arf) inhibitor. Cytotoxic. Antibiotic. Antiviral. Apoptosis inducer. Antitumor compound. Intracellular collagen degradation inhibitor. Reviews.