Learn More
AdipoGen -Brefeldin A
SDP
Catalog No. 502562074
Encompass_Preferred
Chemical. CAS 20350-15-6. Formula C16H24O4. MW 280.4. Isolated from Eupenicillium brefeldianum. Protein transport from ER to Golgi inhibitor. ADP-ribosylation factor Arf inhibitor. Cytotoxic. Antibiotic. Antiviral. Apoptosis inducer. Antitumor compound. Intracellular collagen degradation inhibitor Reviews.
Catalog No. 50-256-2074 Supplier AdipoGen Supplier No. BVT0190M001
Chemical. CAS 20350-15-6. Formula C16H24O4. MW 280.4. Isolated from Eupenicillium brefeldianum. Protein transport from ER to Golgi inhibitor. ADP-ribosylation factor Arf inhibitor. Cytotoxic. Antibiotic. Antiviral. Apoptosis inducer. Antitumor compound. Intracellular collagen degradation inhibitor Reviews.