Learn More
Medchemexpress LLC HY-16592 50mg , Brefeldin A CAS:20350-15-6 Purity:>98%
SDP
Catalog No. 502260190
Encompass_Preferred
HY-16592 50mg Brefeldin A CAS: 20350-15-6 Brefeldin A (BFA) is a lactone antibiotic and a specific inhibitor of protein trafficking. Brefeldin A blocks the transport of secreted and membrane proteins from endoplasmic reticulum to Golgi apparatus. Brefeldin A is also an autophagy and mitophagy inhibitor. Brefeldin A is a CRISPR/Cas9 activator[5]. Brefeldin A inhibits HSV-1 and has anti-cancer activity[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0190 Supplier Medchemexpress LLC Supplier No. HY1659250MG
HY-16592 50mg Brefeldin A CAS: 20350-15-6 Brefeldin A (BFA) is a lactone antibiotic and a specific inhibitor of protein trafficking. Brefeldin A blocks the transport of secreted and membrane proteins from endoplasmic reticulum to Golgi apparatus. Brefeldin A is also an autophagy and mitophagy inhibitor. Brefeldin A is a CRISPR/Cas9 activator[5]. Brefeldin A inhibits HSV-1 and has anti-cancer activity[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.