Learn More
Enzo Life Sciences Brefeldin A, (5mg), CAS Number: 20350-15-6
SDP
Catalog No. NC0652398
Encompass_Preferred
GEF inhibitor. Inhibitor of intracellular protein transport and protein secretion, interfering with trafficking in the .+ channels in epithelia. Induces apoptosis. Alternative Name: BFA, Ascotoxin, Decumbin. Formula: C16H24O4. MW: 280.4. CAS Number: 20350-15-6. MI: 14: 1369. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (20mg/ml), 100% ethanol (5mg/ml), methanol, acetone or ethyl acetate (1mg/ml). Long Term Storage: -20°C
Catalog No. NC0652398 Supplier Enzo Life Sciences Supplier No. BMLG4050005
GEF inhibitor. Inhibitor of intracellular protein transport and protein secretion, interfering with trafficking in the .+ channels in epithelia. Induces apoptosis. Alternative Name: BFA, Ascotoxin, Decumbin. Formula: C16H24O4. MW: 280.4. CAS Number: 20350-15-6. MI: 14: 1369. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (20mg/ml), 100% ethanol (5mg/ml), methanol, acetone or ethyl acetate (1mg/ml). Long Term Storage: -20°C