Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brefeldin A, G-Biosciences
SCI_ED

Catalog No. S139778
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139778 50 mg
S139090 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139778 Supplier G-Biosciences Supplier No. RC903
Add to Cart
Add to Cart

Brefeldin A is an antibiotic derived from Penicillium brefeldianum. It is an anti-cancer agent which disrupts protein transport and causes apoptosis of tumor cells.

Brefeldin A disrupts the function of the Golgi apparatus and blocks Golgi-COPI activity. It has been shown to induce apoptosis in leukemia cells and colon cancer cells.

Chemical Identifiers

Molecular Weight (g/mol) 280.36 g/mol

Specifications

Boiling Point 492.7°C
Quantity 50 mg
Percent Purity 98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.