Learn More
Abcam Brefeldin A, Inhibitor of ADP-ribosylation factor, 25MG
Catalog No. 502707706
Encompass_Preferred
MW 280.36 Da, Purity >98%. Reversible inhibitor of protein translocation from the endoplasmic reticulum to the Golgi apparatus. Inhibits binding of the cytosolic coat protein, β-COP and ADP-ribosylation factor (ARF) to Golgi membranes and inhibits GDP-GTP exchange.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7706 Supplier Abcam Supplier No. AB12029925MG
MW 280.36 Da, Purity >98%. Reversible inhibitor of protein translocation from the endoplasmic reticulum to the Golgi apparatus. Inhibits binding of the cytosolic coat protein, β-COP and ADP-ribosylation factor (ARF) to Golgi membranes and inhibits GDP-GTP exchange.
The product is subject to the following: Abcam Restricted Use Statement