Learn More
Abcam Brefeldin A Solution (1,000 x ), 1ML
Catalog No. NC1081134
Encompass_Preferred
Brefeldin A Solution is supplied as a 1000X in DMSO, which should be diluted to 1X in cell culture medium.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1081134 Supplier Abcam Supplier No. AB1933691ML
Brefeldin A Solution is supplied as a 1000X in DMSO, which should be diluted to 1X in cell culture medium.
The product is subject to the following: Abcam Restricted Use Statement