Learn More
TARGETMOL CHEMICALS INC Bremelanotide Acetate 50MG
SDP
Catalog No. 502876293
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Bremelanotide Acetate (PT-141 Acetate) is an agonist of melanocortin receptor (MCR) . Purity 98%
Catalog No. 50-287-6293 Supplier TARGETMOL CHEMICALS INC Supplier No. T760850MG
Also available in 1 mL, 10 mg, 25 mg, 100 mg and bulk. Please contact Fisher for quotes. Bremelanotide Acetate (PT-141 Acetate) is an agonist of melanocortin receptor (MCR) . Purity 98%