Learn More
Medchemexpress LLC Bremelanotide acetate (PT-141 acetate) | 1607799-13-2 | 100.0% | 1085.2 g/mol | C52H72N14O12 | 10 MG
SDP
Catalog No. 5000294816
Encompass_Preferred
Bremelanotide acetate is a synthetic peptide analogue of α-melanocyte-stimulating hormone that acts as an agonist at melanocortin receptors, primarily MC3R and MC4R. It is supplied as the acetate salt in a lyophilized form for research use, enabling study of melanocortin signaling pathways and sexual function modulation.
  • Acts as an agonist at MC3R and MC4R.
  • Used to study sexual dysfunction and melanocortin signaling.
  • Provided as the acetate salt and lyophilized powder for stability.
  • High purity suitable for research applications.
  • Available in small research quantities, such as 10 mg.
Catalog No. 50-002-94816 Supplier Medchemexpress LLC Supplier No. HY18678A10MG
Bremelanotide acetate is a synthetic peptide analogue of α-melanocyte-stimulating hormone that acts as an agonist at melanocortin receptors, primarily MC3R and MC4R. It is supplied as the acetate salt in a lyophilized form for research use, enabling study of melanocortin signaling pathways and sexual function modulation.
  • Acts as an agonist at MC3R and MC4R.
  • Used to study sexual dysfunction and melanocortin signaling.
  • Provided as the acetate salt and lyophilized powder for stability.
  • High purity suitable for research applications.
  • Available in small research quantities, such as 10 mg.