Learn More
Medchemexpress LLC Bremelanotide acetate (PT-141 acetate) | 1607799-13-2 | 99.81% | 1085.22 | 5 MG
SDP
Catalog No. 5000388141
Encompass_Preferred
Bremelanotide Acetate (PT-141) is a melanocortin receptor agonist that activates MC4R and increases dopamine release. It induces appetitive sexual behaviors, female mounting behavior, and repetitive self-grooming. It can be used for the research of hypoactive sexual desire disorders. For research use only; not for sale to patients.
  • Activates MC4R and increases dopamine release
  • Induces appetitive sexual behaviors
  • Induces female mounting behavior
  • Induces repetitive self-grooming
  • Used for research of hypoactive sexual desire disorders
  • Binds to MC3R and MC4R in a concentration-dependent manner
  • Selectively increases appetitive sexual behaviors in hormone-primed ovariectomized female rats
  • Induces dose-dependent repetitive grooming behavior in male and female mice
Catalog No. 50-003-88141 Supplier Medchemexpress LLC Supplier No. HY18678A5MG
Bremelanotide Acetate (PT-141) is a melanocortin receptor agonist that activates MC4R and increases dopamine release. It induces appetitive sexual behaviors, female mounting behavior, and repetitive self-grooming. It can be used for the research of hypoactive sexual desire disorders. For research use only; not for sale to patients.
  • Activates MC4R and increases dopamine release
  • Induces appetitive sexual behaviors
  • Induces female mounting behavior
  • Induces repetitive self-grooming
  • Used for research of hypoactive sexual desire disorders
  • Binds to MC3R and MC4R in a concentration-dependent manner
  • Selectively increases appetitive sexual behaviors in hormone-primed ovariectomized female rats
  • Induces dose-dependent repetitive grooming behavior in male and female mice