Learn More
Medchemexpress LLC Brensocatib | 1802148-05-5 | 99.9% | 420.46 | C23H24N4O4 | 100 MG
SDP
Catalog No. 5000211104
Encompass_Preferred
Brensocatib is an oral dipeptidyl peptidase 1 (DPP1) inhibitor used in preclinical research to modulate neutrophil serine protease activation. The compound is supplied as a characterized research chemical with documented physicochemical properties and recommended storage conditions.
  • Selective DPP1 inhibition for mechanistic and pharmacology studies.
  • Potent activity reported across species, supporting translational research.
  • Characterized chemical identity with CAS 1802148-05-5 and formula C23H24N4O4.
  • Available in multiple quantities to support assay development and in vitro testing.
  • Storage recommendations provided for powder and solution forms to preserve stability.
Catalog No. 50-002-11104 Supplier Medchemexpress LLC Supplier No. HY101056100MG
Brensocatib is an oral dipeptidyl peptidase 1 (DPP1) inhibitor used in preclinical research to modulate neutrophil serine protease activation. The compound is supplied as a characterized research chemical with documented physicochemical properties and recommended storage conditions.
  • Selective DPP1 inhibition for mechanistic and pharmacology studies.
  • Potent activity reported across species, supporting translational research.
  • Characterized chemical identity with CAS 1802148-05-5 and formula C23H24N4O4.
  • Available in multiple quantities to support assay development and in vitro testing.
  • Storage recommendations provided for powder and solution forms to preserve stability.