Learn More
Medchemexpress LLC Brepocitinib P-Tosylate | 2140301-96-6 | 99.99% | 561.60 | 10 MG
SDP
Catalog No. 5000365038
Encompass_Preferred
Brepocitinib P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor with IC50s of 17 nM and 23 nM, respectively. It also inhibits JAK2 and JAK3 with IC50s of 77 nM and 6.49 μM, respectively. This product is intended for research use only.
  • Potent dual JAK1 and TYK2 inhibitor
  • Inhibits JAK2 and JAK3
  • High purity of 99.99%
  • Available in various quantities and forms
  • Cited in multiple publications
  • Undergoing various clinical trials
Catalog No. 50-003-65038 Supplier Medchemexpress LLC Supplier No. 10MGHY112708A
Brepocitinib P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor with IC50s of 17 nM and 23 nM, respectively. It also inhibits JAK2 and JAK3 with IC50s of 77 nM and 6.49 μM, respectively. This product is intended for research use only.
  • Potent dual JAK1 and TYK2 inhibitor
  • Inhibits JAK2 and JAK3
  • High purity of 99.99%
  • Available in various quantities and forms
  • Cited in multiple publications
  • Undergoing various clinical trials