Learn More
Medchemexpress LLC Brepocitinib (P-Tosylate) | 2140301-96-6 | 99.99% | 561.60 | 100 MG
SDP
Catalog No. 5000364758
Encompass_Preferred
Brepocitinib P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor. It inhibits JAK1 and TYK2 with IC50s of 17 nM and 23 nM, respectively, and also inhibits JAK2 and JAK3. This compound is intended for research use only.
  • Potent dual JAK1 and TYK2 inhibitor
  • Inhibits JAK1 with an IC50 of 17 nM
  • Inhibits TYK2 with an IC50 of 23 nM
  • Inhibits JAK2 and JAK3
  • Applicable for in vitro and in vivo research
Catalog No. 50-003-64758 Supplier Medchemexpress LLC Supplier No. 100MGHY112708A
Brepocitinib P-Tosylate is a potent dual Janus kinase 1 (JAK1) and TYK2 inhibitor. It inhibits JAK1 and TYK2 with IC50s of 17 nM and 23 nM, respectively, and also inhibits JAK2 and JAK3. This compound is intended for research use only.
  • Potent dual JAK1 and TYK2 inhibitor
  • Inhibits JAK1 with an IC50 of 17 nM
  • Inhibits TYK2 with an IC50 of 23 nM
  • Inhibits JAK2 and JAK3
  • Applicable for in vitro and in vivo research