Learn More
Medchemexpress LLC 6-fluoro-2-[4-(2-fluorophenyl)phenyl]-3-methylquinoline-4-carboxylic acid | 96187-53-0 | MFCD00866437 | 99.9% | 375.37 | C23H15F2NO2 | 10 MG
SDP
Catalog No. 501968528
Encompass_Preferred
Brequinar is a small-molecule inhibitor of dihydroorotate dehydrogenase (DHODH) used as a research reagent. It shows potent DHODH inhibition (human DHODH IC50 ≈ 5.2 nM) and has reported antiviral activity in preclinical studies. Supplied as a solid for preparing stock solutions, it is intended for laboratory research use only.
  • Potent DHODH inhibitor with reported human IC50 ≈ 5.2 nM.
  • High purity as supplied (99.9%).
  • Molecular formula C23H15F2NO2 and molecular weight 375.37 g/mol.
  • Supplied as a solid suitable for preparing stock solutions.
  • Applicable to antiviral and metabolic pathway research.
  • For research use only; not for human or diagnostic use.
Catalog No. 50-196-8528 Supplier Medchemexpress LLC Supplier No. HY10832510MG
Brequinar is a small-molecule inhibitor of dihydroorotate dehydrogenase (DHODH) used as a research reagent. It shows potent DHODH inhibition (human DHODH IC50 ≈ 5.2 nM) and has reported antiviral activity in preclinical studies. Supplied as a solid for preparing stock solutions, it is intended for laboratory research use only.
  • Potent DHODH inhibitor with reported human IC50 ≈ 5.2 nM.
  • High purity as supplied (99.9%).
  • Molecular formula C23H15F2NO2 and molecular weight 375.37 g/mol.
  • Supplied as a solid suitable for preparing stock solutions.
  • Applicable to antiviral and metabolic pathway research.
  • For research use only; not for human or diagnostic use.