Learn More
TARGETMOL CHEMICALS INC BREQUINAR 25MG
SDP
Catalog No. 502579823
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Brequinar (NSC-368390) is a potent inhibitor of dihydroorotate dehydrogenase with potent activities against a broad spectrum of viruses. purity: 99%
Catalog No. 50-257-9823 Supplier TARGETMOL CHEMICALS INC Supplier No. T501325MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Brequinar (NSC-368390) is a potent inhibitor of dihydroorotate dehydrogenase with potent activities against a broad spectrum of viruses. purity: 99%