Learn More
Cayman Chemical Brequinar (sodium salt), Purity: ≥98%, CAS: 96201-88-6, 50 mg
SDP
Catalog No. 502911865
Encompass_Preferred
A DHODH inhibitor (IC50 = ~20 nM); selective for DHODH over a panel of greater than 400 kinases at 100 nM; induces differentiation of ER-HoxA9, U937, and THP-1 cells in vitro (EC50s = ~1 µM); induces differentiation of tumor cells and reduces tumor growth in a THP-1 mouse xenograft model at 15 mg/kg three times daily or 5 mg/kg per day; induces differentiation of bone marrow leukemic cells and increases survival In a retroviral transduction mouse model of HoxA9 + Meis1 AML
Catalog No. 50-291-1865 Supplier Cayman Chemical Supplier No. 3618350 MG
A DHODH inhibitor (IC50 = ~20 nM); selective for DHODH over a panel of greater than 400 kinases at 100 nM; induces differentiation of ER-HoxA9, U937, and THP-1 cells in vitro (EC50s = ~1 µM); induces differentiation of tumor cells and reduces tumor growth in a THP-1 mouse xenograft model at 15 mg/kg three times daily or 5 mg/kg per day; induces differentiation of bone marrow leukemic cells and increases survival In a retroviral transduction mouse model of HoxA9 + Meis1 AML