Learn More
Strem, An Ascensus Company CAS# 1070663-78-3. 2-(Dicyclohexylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% BrettPhos. MFCD11973797. 25G
SDP
Supplier:  Strem, An Ascensus Company 15115225G
Encompass
CAS# 1070663-78-3. 25G. 2-(Dicyclohexylphosphino)-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl, min. 98% BrettPhos. MFCD11973797. Molecular Formula: C35H53O2P. Molecular Weight: 536.77. Color/Form: white xtl. Strem#: 15-1152 www.strem.com/catalog/v3/15-1152/
Catalog No. 50-195-9518