Learn More
Strem, An Ascensus Company CAS# 1599466-83-7. Methanesulfonato(2-dicyclohexylphosphino-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II), min. 98% MFCD28411654. 250MG
SDP
Supplier:  Strem, An Ascensus Company 460333250MG
Encompass
CAS# 1599466-83-7. 250MG. Methanesulfonato(2-dicyclohexylphosphino-3,6-dimethoxy-2',4',6'-tri-i-propyl-1,1'-biphenyl)(2'-methylamino-1,1'-biphenyl-2-yl)palladium(II), min. 98% [BrettPhos Palladacycle Gen. 4]. MFCD28411654. Molecular Formula: C49H68NO5PPdS. Molecular Weight: 920.53. Color/Form: off-white to beige pwdr. Strem#: 46-0333. www.strem.com/catalog/v3/46-0333/
Catalog No. 50-195-9478