Learn More
Medchemexpress LLC Bretylium tosylate | 61-75-6 | 99.9% | 10 MG
SDP
Catalog No. 5000340715
Encompass_Preferred
Bretylium (tosylate) is an inhibitor of the presynaptic release of vasoconstrictor neurotransmitters. It acts as a sympathetic nerve and adrenergic ganglion blocking agent. It inhibits adrenergic function presynaptically only after an initial release in neurotransmitter substance.
  • Inhibitor of presynaptic release of vasoconstrictor neurotransmitters.
  • Acts as a sympathetic nerve and adrenergic ganglion blocking agent.
  • Inhibits adrenergic function presynaptically.
Catalog No. 50-003-40715 Supplier Medchemexpress LLC Supplier No. HY12961A10MG
Bretylium (tosylate) is an inhibitor of the presynaptic release of vasoconstrictor neurotransmitters. It acts as a sympathetic nerve and adrenergic ganglion blocking agent. It inhibits adrenergic function presynaptically only after an initial release in neurotransmitter substance.
  • Inhibitor of presynaptic release of vasoconstrictor neurotransmitters.
  • Acts as a sympathetic nerve and adrenergic ganglion blocking agent.
  • Inhibits adrenergic function presynaptically.