Learn More
Cayman Chemical Bretylium (tosylate), Purity: ≥95%, CAS: 61-75-6, 10 mg
SDP
Catalog No. 502989706
Encompass_Preferred
A class III antiarrhythmic agent; has antiadrenergic activity in isolated tissue preparations; an inhibitor of the Na+/K+-ATPase (IC50 = 4.5 mM); prevents ventricular fibrillation in anesthetized dogs in a model of sudden coronary death at 10 mg/kg; inhibits NCTs, as well as increases action potential delay and the absolute refractory period in isolated mouse vas deferens sympathetic nerve terminals
Catalog No. 50-298-9706 Supplier Cayman Chemical Supplier No. 2997910 MG
A class III antiarrhythmic agent; has antiadrenergic activity in isolated tissue preparations; an inhibitor of the Na+/K+-ATPase (IC50 = 4.5 mM); prevents ventricular fibrillation in anesthetized dogs in a model of sudden coronary death at 10 mg/kg; inhibits NCTs, as well as increases action potential delay and the absolute refractory period in isolated mouse vas deferens sympathetic nerve terminals