Learn More
ATCC Brevibacillus agri
Catalog No. 502383285
Encompass
Brevibacillus agri strain NRS 1219 is a bacterial type strain that is included in the bioMérieux VITEK 2 - Bacillus (BCL) Card Quality Control Organism Set.
Catalog No. 50-238-3285 Supplier ATCC Supplier No. 51663
Brevibacillus agri strain NRS 1219 is a bacterial type strain that is included in the bioMérieux VITEK 2 - Bacillus (BCL) Card Quality Control Organism Set.