Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Brevibacillus agri derived from ATCC 51663, KWIK-STIK Plus
SDP

Catalog No. 23047246 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
5 KWIK-STIKS
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-246 5 KWIK-STIKS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-246 Supplier Microbiologics™ Supplier No. 0139X
Add to Cart
Add to Cart
This item is not returnable. View return policy

KWIK-STIK Plus combines microorganisms that are two passages from reference strains, with KWIK-STIK format. Each device features a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab. Everything needed for growing control cultures is included in one device.

  • Biosafety Level: 1
  • 5 KWIK-STIK™ units with a microorganism limited to 2 passages from the reference culture.

Specifications

ATCC® ATCC 51663
Biosafety Level 1
Content And Storage 2°C to 8°C
Format KWIK-STIK Plus
Manufacturing Origin United States
Product Type Quality Control Organism
For Use With (Application) Microbial Identification
Packaging Type Plastic Canister
Quantity 5 KWIK-STIKS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.