Learn More
Medchemexpress LLC HY-N4095 5mg Medchemexpress, Brevifolincarboxylic acid CAS:18490-95-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN40955MG
Encompass_Preferred
Medchemexpress, HY-N4095 5mg Brevifolincarboxylic acid CAS:18490-95-4 Brevifolincarboxylic acid is extracted from Polygonum capitatum, has inhibitory effect on the aryl hydrocarbon receptor (AhR). Brevifolincarboxylic acid is an α-glucosidase inhibitor with an IC50 of 323.46 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2922