Learn More
Cayman Chemical Brevifolincarboxylic Acid, Purity: ≥95%, CAS: 18490-95-4, 10 mg
SDP
Catalog No. 502910356
Encompass_Preferred
A phenol with anticancer and anti-inflammatory activities; inhibits the proliferation of PC-14 lung cancer cells (IC50 = 3.95 µg/ml); reduces LPS-induced increases in NO production in RAW 264.7 macrophages at 20 µM
Catalog No. 50-291-0356 Supplier Cayman Chemical Supplier No. 3492210 MG
A phenol with anticancer and anti-inflammatory activities; inhibits the proliferation of PC-14 lung cancer cells (IC50 = 3.95 µg/ml); reduces LPS-induced increases in NO production in RAW 264.7 macrophages at 20 µM