Learn More
Medchemexpress LLC Brevilin A | 16503-32-5 | 1 MG
SDP
Catalog No. 5000421323
Encompass_Preferred
Brevilin A is an orally active STAT3/JAK inhibitor with anti-tumor and anti-proliferative activities against cancer cells. It also induces apoptosis and autophagy, and inhibits STAT3 signaling.
  • Orally active STAT3/JAK inhibitor
  • Demonstrates anti-tumor activity
  • Exhibits anti-proliferative activity against cancer cells
  • Induces apoptosis and autophagy
  • Inhibits STAT3 signaling in a dose-dependent manner
  • Blocks STAT3 tyrosine 705 phosphorylation
  • Inhibits CNE-2 xenograft tumor growth and PI3K/AKT and STAT3 signaling in vivo
Catalog No. 50-004-21323 Supplier Medchemexpress LLC Supplier No. HYN29591MG
Brevilin A is an orally active STAT3/JAK inhibitor with anti-tumor and anti-proliferative activities against cancer cells. It also induces apoptosis and autophagy, and inhibits STAT3 signaling.
  • Orally active STAT3/JAK inhibitor
  • Demonstrates anti-tumor activity
  • Exhibits anti-proliferative activity against cancer cells
  • Induces apoptosis and autophagy
  • Inhibits STAT3 signaling in a dose-dependent manner
  • Blocks STAT3 tyrosine 705 phosphorylation
  • Inhibits CNE-2 xenograft tumor growth and PI3K/AKT and STAT3 signaling in vivo