Learn More
TARGETMOL CHEMICALS INC BREVILIN A 5MG
SDP
Catalog No. 502579855
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Brevilin A is a sesquiterpene lactone isolated from Centipeda minima inhibits janus kinase activity and blocks STAT3 signaling in cancer cells with anti-tumor activity. Brevilin A is a selective inhibitor of JAK-STAT signal pathway by attenuating the JAKs activity and blocking STAT3 signaling (IC50 = 10.6 uM) in Cancer Cells.Brevilin A induces apoptosis and autophagy via mitochondrial pathway and PI3K/AKT/mTOR inactivation in colon adenocarcinoma cell CT26.antitumourpurity: 99%
Catalog No. 50-257-9855 Supplier TARGETMOL CHEMICALS INC Supplier No. T46725MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Brevilin A is a sesquiterpene lactone isolated from Centipeda minima inhibits janus kinase activity and blocks STAT3 signaling in cancer cells with anti-tumor activity. Brevilin A is a selective inhibitor of JAK-STAT signal pathway by attenuating the JAKs activity and blocking STAT3 signaling (IC50 = 10.6 uM) in Cancer Cells.Brevilin A induces apoptosis and autophagy via mitochondrial pathway and PI3K/AKT/mTOR inactivation in colon adenocarcinoma cell CT26.antitumourpurity: 99%