Learn More
Medchemexpress LLC HY-N2959 5mg Medchemexpress, Brevilin A CAS:16503-32-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN29595MG
Encompass_Preferred
Medchemexpress, HY-N2959 5mg Brevilin A CAS:16503-32-5 Brevilin A is a sesquiterpene lactone isolated from Centipeda minima with anti-tumor activity. Brevilin A is a selective inhibitor of JAK-STAT signal pathway by attenuating the JAKs activity and blocking STAT3 signaling (IC50 = 10.6 µM) in Cancer Cells. Brevilin A induces apoptosis and autophagy via mitochondrial pathway and PI3K/AKT/mTOR inactivation in colon adenocarcinoma cell CT26. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0407