Learn More
ATCC Brevundimonas diminuta strain PCI 818 (ATCC 19146™)
Supplier:  ATCC 19146
Encompass
Brevundimonas diminuta strain PCI 818 (ATCC 19146™) is a bacterial strain used in pharmaceutical and personal care, filtration sterilization control, membrane filter testing, and sterility assurance.
Catalog No. 50-238-2409