Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxoid Culti-Loops™ Brevundimonas dimunita ATCC™ 19146™, 5 Pk

Catalog No. R4607200 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 Loops/Pk
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R4607200 5 Loops/Pk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R4607200 Supplier Oxoid Supplier No. R4607200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.

The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Cat. No. R4631000 or streaked directly onto an appropriate media.

Ordering Information:

Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2 to 8°C
Format Culti-Loops
Product Type Quality Control Organism
Target Organism Class Bacteria
For Use With (Application) Bioinformatics, Quality control, Pharmaceutical testing
Quantity 5 Loops/Pk
Strain Designations FDA strain PCI 818 [CCUG 24715, DSM 1635, LMG 10743]
Source Contaminant in culture of Bacillus cereus
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.