Learn More
Brew Plate™ Blk Yeast Anaerobic Plate
SCI_ED
Catalog No. S43556
Change view
Click to view available options
Packaging Type:
Bulk Sleeve
Catalog No. Packaging Type
S43556 Bulk Sleeve
1 options
Catalog No. S43556 Supplier Bioplast Manufacturing (Brew Plate) Supplier No. BREWBULKYAQP
For detection of all yeast and bacterial contamination found in brew.
  • Each quadrant made from different media
  • Contains Lins cupric sulfate media, Lee multi-dimenstional agar, Lins Wild Yeast Media and Wallerstein Nutrient Media
Specifications
Media Type Lin’s Cupric Sulfate Media, Lin’s Wild Yeast Media, Wallerstein Nutrient Media
Product Type Microbiology Media
Packaging Type Bulk Sleeve