Learn More
LaMotte™ BrewLab™ Test Kit
SDPSCI_ED
Catalog No. S43549 Shop All LaMotte Company Products
Change view
Click to view available options
Includes:
Tests for 11 water quality factors
Tests for 7 water quality factors
Tests for 8 water quality factors; Digital pH meter
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S43549 Tests for 11 water quality factors
S43551 Tests for 7 water quality factors
S43550 Tests for 8 water quality factors; Digital pH meter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S43549 Supplier LaMotte™ Supplier No. 7190
Add to Cart
Add to Cart
LaMotte™ BrewLab™ is an economical, easy-to-use test kit to measure the mineral content of water by monitoring essential water quality factors for better home-brewed beer.
The quality of the water that you start with will affect your final product, so take the mystery out of making great-tasting beer with the BrewLab™ Test Kit for home brewers.
  • Economical, easy-to-use test kit to quickly measure the mineral content of water and allow the brewer to make informed decisions in the brewing process
  • Alkalinity is the carbonate and bicarbonate content of the water, which acts to raise the pH of the mash and beer
  • Chloride ion acts to bring out the sweetness and fullness of the malt flavor
  • Dissolved oxygen can have both a positive and a negative impact on beer
  • Fermentation itself is an anerobic process, but yeast cells require oxygen for growth
  • Calcium helps prevent enzymes from denaturing and extends their activity in mash, promoting clarity, flavor and stability
  • Magnesium is an important yeast nutrient
  • The mineral content of the water directly affects the pH of the mash, and the pH of the mash affects the entire brewing process
  • Sodium acts in concert with chloride to enhance the sweetness and fullness of the malt, but too much can oversalt the beer and result in salty or metallic flavors
  • Sulfate ion acts to accenturate the hoppiness and dryness of the beer, making it more crisp tasting
  • Measurement of total dissolved solids allows the brewer to quickly determine if there has been a shift or change in a water source or brewing process as well as provide a QA check on demineralization and waste treatment effectiveness
Specifications
For Use With (Application) Quantifies 11 Water Quality Factors
Includes Tests for 11 water quality factors
Product Type BrewLab Pro
WARNING: Cancer - www.P65Warnings.ca.gov