Learn More
Aviva Systems Biology Corp BRF1 Antibody - C-terminal region (ARP37387_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37387_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against BRF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: GTF3B, TAF3C, TAFIII90, TFIIIB90, mTFIIIB90, 2510002F24RikApplicationWB. Species reactivity: Cow, Dog, Mouse, Rat, Yeast
Catalog No. 50-014-56359