Learn More
Aviva Systems Biology Corp BRF1 Antibody - C-terminal region (ARP40137_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40137_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of mouse BRF1. Alternate names: GTF3B, TAF3C, TAFIII90, TFIIIB90, mTFIIIB90, 2510002F24RikApplicationWB. Species reactivity: Guinea Pig, Mouse, Rabbit, Rat
Catalog No. 50-014-57305