Learn More
Aviva Systems Biology Corp BRF2 Antibody - middle region (ARP50631_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50631_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRF2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human BRF2. Alternate names: BRFU, FLJ11052, TFIIIB50ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-523