Learn More
ATHENA ES BRFF-HPC1
Catalog No. NC9970798
Encompass_Preferred
BRFF-HPC-1™ is a complete serum-free medium comprised of essential amino acids, vitamins, inorganic salts, trace elements, growth factors and dihydrotestosterone, designed for establishing new cell lines from human prostate tissue. HPC-1™ was developed and has been extensively used for establishing and maintaining epithelial cell lines from both benign prostatic hyperplasia and prostatic carcinoma. Optimal attached monolayer cell growth can be achieved when used in conjunction with FNC Coating Mix.
Catalog No. NC9970798 Supplier ATHENA ES Supplier No. 0403
BRFF-HPC-1™ is a complete serum-free medium comprised of essential amino acids, vitamins, inorganic salts, trace elements, growth factors and dihydrotestosterone, designed for establishing new cell lines from human prostate tissue. HPC-1™ was developed and has been extensively used for establishing and maintaining epithelial cell lines from both benign prostatic hyperplasia and prostatic carcinoma. Optimal attached monolayer cell growth can be achieved when used in conjunction with FNC Coating Mix.