Learn More
EPICYPHER INC BRG1/SMARCA4 CUTANA™ CUT&RUN Antibody, 100 µL
SDP
Catalog No. NC2944936
Encompass
This antibody meets EpiCypher’s “CUTANA Compatible” criteria for performance in Cleavage Under Targets and Release Using Nuclease (CUT&RUN) and/or Cleavage Under Targets and Tagmentation (CUT&Tag) approaches to genomic mapping. Every lot of a CUTANA Compatible antibody is tested in the indicated CUTANA approach using EpiCypher optimized protocols and determined to yield peaks that show a genomic distribution pattern consistent with reported function(s) of the target protein. BRG1 antibody produces CUT&RUN peaks primarily flanking transcription start sites (TSSs, Figure 1). BRG1 peaks show a large degree of overlap with BRD4 peaks (Figure 2), as has been reported in the literature [Conrad et al., 2017].
Catalog No. NC2944936 Supplier EPICYPHER INC Supplier No. 132002
This antibody meets EpiCypher’s “CUTANA Compatible” criteria for performance in Cleavage Under Targets and Release Using Nuclease (CUT&RUN) and/or Cleavage Under Targets and Tagmentation (CUT&Tag) approaches to genomic mapping. Every lot of a CUTANA Compatible antibody is tested in the indicated CUTANA approach using EpiCypher optimized protocols and determined to yield peaks that show a genomic distribution pattern consistent with reported function(s) of the target protein. BRG1 antibody produces CUT&RUN peaks primarily flanking transcription start sites (TSSs, Figure 1). BRG1 peaks show a large degree of overlap with BRD4 peaks (Figure 2), as has been reported in the literature [Conrad et al., 2017].