Learn More
Aviva Systems Biology Corp BRI3BP Antibody - C-terminal region (ARP50260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP50260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRI3BP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human BRI3BP. Alternate names: BNAS1, HCCR-2, KG19, HCCR-1, HCCRBP-1ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-60302