Learn More
TARGETMOL CHEMICALS INC BRICICLIB 10MG
SDP
Catalog No. 502576976
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Briciclib (ON 014185) is a small molecule that suppresses cyclin D1 accumulation in Y cells. purity: 99%
Catalog No. 50-257-6976 Supplier TARGETMOL CHEMICALS INC Supplier No. T320710MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Briciclib (ON 014185) is a small molecule that suppresses cyclin D1 accumulation in Y cells. purity: 99%