Learn More
Selleck Chemical LLC Briciclib
SDP
Supplier:  Selleck Chemical LLC S65332MG
Encompass_Preferred
Briciclib (ON-014185) is a water soluble derivative of ON-013100 a small molecule that binds to and inhibits eukaryotic translation initiation factor 4E (eIF4E)
Catalog No. 50-000-02920