Learn More
Plastilite Corp Hot and Cold Packs. Refrigerant Ice Pack. Foam Brick Ice Pack. ControliQ foam brick pack from Integritemp/Plastilite. Freeze and use as wet ice replacement in chilled and frozen product shipments. #TFB18-32 18oz. 7.125"x5.125"x.875” 32/case
Catalog No. NC3579785
Encompass_Preferred
Hot and Cold Packs. Refrigerant Ice Pack. Foam Brick Ice Pack. ControliQ foam brick pack from Integritemp/Plastilite. Freeze and use as wet ice replacement in chilled and frozen product shipments. #TFB18-32, 18oz. 7.125"x5.125"x.875” 32/case ControliQ Foam Brick Packs offer dimensional consistency for easy packing from one insulated package to the next. This consistency is vital when packages are designed with tight tolerances for product payload and refrigerant packs. They are non-toxic and reusable. Packaged in heavy duty vented corrugated boxes for secure shipping & storage and accelerated freezing.
Catalog No. NC3579785 Supplier Plastilite Corp Supplier No. TFB1832
Hot and Cold Packs. Refrigerant Ice Pack. Foam Brick Ice Pack. ControliQ foam brick pack from Integritemp/Plastilite. Freeze and use as wet ice replacement in chilled and frozen product shipments. #TFB18-32, 18oz. 7.125"x5.125"x.875” 32/case ControliQ Foam Brick Packs offer dimensional consistency for easy packing from one insulated package to the next. This consistency is vital when packages are designed with tight tolerances for product payload and refrigerant packs. They are non-toxic and reusable. Packaged in heavy duty vented corrugated boxes for secure shipping & storage and accelerated freezing.