Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bridge Building Class Pack
SCI_ED

Catalog No. S41893B
Change view
Click to view available options
Product Type:
Kit, Balsa Bridge Building
Kit, Bass Bridge Building
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S41893B Kit, Balsa Bridge Building
S41893A Kit, Bass Bridge Building
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S41893B Supplier Saunders Midwest LLC Supplier No. 8660
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

All materials required:

  • Wood
  • Adhesive
  • Competition design parameters
  • Booklet: Teaching with Model Bridge Building Programs

Challenge students' mathematical skills with this analytical exercise in creativity.

  • Supports a wide variety of curricula including geometry, physics, technology, mathematics and general science
  • Build the strongest bridge with the smallest amount of materials
  • Bridge efficiency is judged as the ratio of the weight held by the bridge to the weight of the bridge

Order Info

Select 360 pieces of either 0.13 x 0.13 x 24 in. (0.31 x 0.31 x 60cm) balsa wood or 0.14 x 0.14 x 24 in. (0.34 x 0.34 x 60cm) basswood; Class size: 24 students

Specifications

Product Type Kit, Balsa Bridge Building
Grade 6 to 12
For Use With (Application) For student's mathematical skills with this analytical exercise in creativity
Includes Wood, adhesive, competition design parameters, booklet: “teaching with model bridge building programs”?
Quantity Supplies 24 students
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.