Learn More
World Precision Instrument Bridge8 Transducer Amplifier Module
SDP
Catalog No. 502521211
Encompass
Low noise transducer amplifier in a modular system
Bridge8 is a modular, rack-mountable amplifier system. It is specifically designed for use as a signal conditioning amplifier with strain gages and other poered transducers. Bridge8 includes differential amplifiers featuring high input impedance, high common mode rejection and low current leakage input terminals for low noise operation. The Bridge8 also features a half bridge switch and channel offset. A wide variety of WPI transducers are available for force, temperature, pressure and light measurements. The Bridge8 amplifier is a clear choice for convenience and quality.
Any combination of ISO-DAM8 or Bridge8 modules may be combined in the same enclosure. The ISO-DAM8A module is also available in this rack configuration.
Catalog No. 50-252-1211 Supplier World Precision Instrument Supplier No. BRIDGE8
Low noise transducer amplifier in a modular system
Bridge8 is a modular, rack-mountable amplifier system. It is specifically designed for use as a signal conditioning amplifier with strain gages and other poered transducers. Bridge8 includes differential amplifiers featuring high input impedance, high common mode rejection and low current leakage input terminals for low noise operation. The Bridge8 also features a half bridge switch and channel offset. A wide variety of WPI transducers are available for force, temperature, pressure and light measurements. The Bridge8 amplifier is a clear choice for convenience and quality.
Any combination of ISO-DAM8 or Bridge8 modules may be combined in the same enclosure. The ISO-DAM8A module is also available in this rack configuration.