Learn More
Medchemexpress LLC HY-12857 50mg , Brigatinib CAS:1197953-54-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1285750MG
Encompass_Preferred
HY-12857 50mg Brigatinib CAS: 1197953-54-0 Brigatinib (AP-26113) is a highly potent and selective ALK inhibitor, with an IC50 of 0.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0661